2-Amino-4-methylthiazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methylthiazol-2-ylamine, 4-Methylthiazol-2-amine, 2-Amino-4-methylthiazol

HI-1202-25G HI-1202-100G
COBBHI-1202-25GEA 0 GBP
COBBHI-1202-25G COBBHI-1202-100G
2-Amino-4-methylthiazole
2-Amino-4-methylthiazole
Formula: C₄H₆N₂S
MW: 114.17 g/mol
Boiling Pt: 231 °C (1013 hPa)
Melting Pt: 42 °C
Density: 1.202 g/cm³ (50 °C)
MDL Number: MFCD00005329
CAS Number: 1603-91-4
EINECS: 216-505-6
Merck Index: 13,00449

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR