2-Amino-3-bromo-5-methylbenzoic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

631531-1G 631531-5G
SIAL631531-1GEA 62.5 GBP
SIAL631531-1G SIAL631531-5G
2-Amino-3-bromo-5-methylbenzoic acid, Sigma-Aldrich®
2-Amino-3-bromo-5-methylbenzoic acid
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD00051705
CAS Number: 13091-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR