2-Amino-3,5-dibromopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 3,5-Dibromo-2-pyridylamine, 3,5-Dibromopyridin-2-amine

PY-7080-100G PY-7080-25G PY-7080-5G
COBBPY-7080-100GEA 0 GBP
COBBPY-7080-100G COBBPY-7080-25G COBBPY-7080-5G
2-Amino-3,5-dibromopyridine
2-Amino-3,5-dibromopyridine
Formula: C₅H₄Br₂N₂
MW: 251.91 g/mol
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 104 °C
MDL Number: MFCD00038041
CAS Number: 35486-42-1
EINECS: 252-590-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR