2,6-Difluoro-3-methylbenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,6-Difluoro-3-tolualdehyde, 2,6-Difluoro-m-tolualdehyde, 2,6-Difluoro-m-tolylaldehyde, 2,6-Difluoro-3-tolylaldehyde

005747-5G 005747-1G
FLUO005747-5GEA 0 GBP
FLUO005747-5G FLUO005747-1G
2,6-Difluoro-3-methylbenzaldehyde
2,6-Difluoro-3-methylbenzaldehyde
Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD01631333
CAS Number: 261763-34-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR