2,5-Dichlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2662-25G BB-2662-1G BB-2662-5G
COBBBB-2662-25GEA 0 GBP
COBBBB-2662-25G COBBBB-2662-1G COBBBB-2662-5G
2,5-Dichlorophenylboronic acid
2,5-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
MDL Number: MFCD01863182
CAS Number: 135145-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR