2,5-Dibromo-4-methylimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,5-Dibromo-4-methyl-1H-imidazole

HI-1032-1G HI-1032-5G
COBBHI-1032-1GEA 0 GBP
COBBHI-1032-1G COBBHI-1032-5G
2,5-Dibromo-4-methylimidazole
2,5-Dibromo-4-methylimidazole
Formula: C₄H₄Br₂N₂
MDL Number: MFCD02179519
CAS Number: 219814-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR