2,4-Difluoropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7534-25G OS-7534-1G OS-7534-5G
COBBOS-7534-25GEA 0 GBP
COBBOS-7534-25G COBBOS-7534-1G COBBOS-7534-5G
2,4-Difluoropyridine
2,4-Difluoropyridine
Formula: C₅H₃F₂N
MDL Number: MFCD04114162
CAS Number: 34941-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR