2,4-Bis(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3518-5G BB-3518-25G BB-3518-1G
COBBBB-3518-5GEA 0 GBP
COBBBB-3518-5G COBBBB-3518-25G COBBBB-3518-1G
2,4-Bis(trifluoromethyl)phenylboronic acid
2,4-Bis(trifluoromethyl)phenylboronic acid
Formula: C₈H₅BF₆O₂
MW: 257.93 g/mol
MDL Number: MFCD01631349
CAS Number: 153254-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR