2,4-Bis(trifluoromethyl)benzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004999-5G 004999-1G
FLUO004999-5GEA 0 GBP
FLUO004999-5G FLUO004999-1G
2,4-Bis(trifluoromethyl)benzamide
2,4-Bis(trifluoromethyl)benzamide
Formula: C₉H₅F₆NO
MW: 257.14 g/mol
MDL Number: MFCD00083536
CAS Number: 53130-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR