2,4,6-Trifluorobenzaldehyde
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7717-5G
OS-7717-1G
COBBOS-7717-5GEA
0
GBP
COBBOS-7717-5G
COBBOS-7717-1G
2,4,6-Trifluorobenzaldehyde
2,4,6-Trifluorobenzaldehyde
Formula:
C₇H₃F₃O MW: 160.1 g/mol |
MDL Number:
MFCD00061197 CAS Number: 58551-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...