2,3-Difluoro-4-methylbenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3-Difluoro-p-tolualdehyde, 2,3-Difluoro-p-tolylaldehyde, 2,3-Difluoro-4-tolylaldehyde, 2,3-Difluoro-4-tolualdehyde

005746-5G 005746-1G
FLUO005746-5GEA 0 GBP
FLUO005746-5G FLUO005746-1G
2,3-Difluoro-4-methylbenzaldehyde
2,3-Difluoro-4-methylbenzaldehyde
Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD01631644
CAS Number: 245536-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR