2,3-Dibromopyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
PY-1458-1G
PY-1458-25G
PY-1458-5G
COBBPY-1458-1GEA
0
GBP
COBBPY-1458-1G
COBBPY-1458-25G
COBBPY-1458-5G
2,3-Dibromopyridine
2,3-Dibromopyridine
Formula:
C₅H₃Br₂N MW: 236.89 g/mol Boiling Pt: 249…249.5 °C (1013 hPa) Melting Pt: 58…59 °C |
MDL Number:
MFCD00234014 CAS Number: 13534-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...