2,3,4-Trifluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3,4-Trifluorobenzeneboronic acid, Dihydroxy(2,3,4-trifluorophenyl)borane, (2,3,4-Trifluorophenyl)metaboric acid

BB-3519-25G BB-3519-1G BB-3519-100G BB-3519-5G
COBBBB-3519-25GEA 0 GBP
COBBBB-3519-25G COBBBB-3519-1G COBBBB-3519-100G COBBBB-3519-5G
2,3,4-Trifluorophenylboronic acid
2,3,4-Trifluorophenylboronic acid
Formula: C₆H₄BF₃O₂
MW: 175.9 g/mol
MDL Number: MFCD01863168
CAS Number: 226396-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR