Thiomersal

Supplier: EHRENSTORFER GMBH
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-(Ethylmercuriomercapto)benzoic acid sodium saltEthylmercurithiosalicylic acid sodium saltThimerosalMerthiolat2-[(Ethylmercury)thio]benzoic acid sodium saltSodium ethylmercurithiosalicylate2-(Ethylmercuriothio)benzoic acid sodium salt

EHERC17515000EA 397 GBP
EHERC17515000
Thiomersal
Thiomersal
Formula: 2-(C₂H₅HgS)C₆H₄CO₂Na
MW: 404.82 g/mol
Melting Pt: 232…233 °C
Flash Pt: 250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00013062
CAS Number: 54-64-8
EINECS: 200-210-4
UN: 2025
ADR: 6.1,II
Merck Index: 12,09451
https://uk.vwr.com/stibo/web/std.lang.all/72/11/2297211.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR