1,2,3-Trifluoro-4-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3,4-Trifluoronitrobenzene

BK-4510-25G BK-4510-1KG BK-4510-100G
COBBBK-4510-25GEA 0 GBP
COBBBK-4510-25G COBBBK-4510-1KG COBBBK-4510-100G
1,2,3-Trifluoro-4-nitrobenzene
1,2,3-Trifluoro-4-nitrobenzene
Formula: C₆H₂F₃NO₂
MW: 177.08 g/mol
Boiling Pt: 92 °C (1013 hPa)
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00041546
CAS Number: 771-69-7
EINECS: 212-238-4
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR