3-(Trifluoromethyl)anisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OT-0720-25G OT-0720-100G
COBBOT-0720-25GEA 0 GBP
COBBOT-0720-25G COBBOT-0720-100G
3-(Trifluoromethyl)anisole
3-(Trifluoromethyl)anisole
Formula: C₈H₇F₃O
MW: 176.14 g/mol
Boiling Pt: 159.5 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1.2426 g/cm³ (20 °C)
MDL Number: MFCD00000385
CAS Number: 454-90-0
EINECS: 207-229-7
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR