4-Fluoro-2-methoxybenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013332-5G 013332-1G 013332-25G
FLUO013332-5GEA 0 GBP
FLUO013332-5G FLUO013332-1G FLUO013332-25G
4-Fluoro-2-methoxybenzonitrile
4-Fluoro-2-methoxybenzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD04116335
CAS Number: 191014-55-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR