5-Fluoro-2-methoxybenzonitrile, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Cyano-4-fluoroanisole

M-Clarity™ quality level = MQ100

527734-1G 527734-5G
SIAL527734-1GEA 52.2 GBP
SIAL527734-1G SIAL527734-5G
5-Fluoro-2-methoxybenzonitrile, Sigma-Aldrich®
5-Fluoro-2-methoxybenzonitrile
Formula: C₈H₆FNO
MW: 151.14 g/mol
MDL Number: MFCD02683503
CAS Number: 189628-38-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR