3-Fluoro-4-hydroxybenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013830-25G 013830-5G
FLUO013830-25GEA 0 GBP
FLUO013830-25G FLUO013830-5G
3-Fluoro-4-hydroxybenzonitrile
3-Fluoro-4-hydroxybenzonitrile
Formula: C₇H₄FNO
MW: 137.11 g/mol
MDL Number: MFCD00215834
CAS Number: 405-04-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR