2-Fluoro-6-iodobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1054-5G AN-1054-25G AN-1054-100G
COBBAN-1054-5GEA 7.91 GBP
COBBAN-1054-5G COBBAN-1054-25G COBBAN-1054-100G
2-Fluoro-6-iodobenzonitrile
2-Fluoro-6-iodobenzonitrile
Formula: C₇H₃FIN
MW: 247.01 g/mol
Boiling Pt: 281 °C (1013 hPa)
Melting Pt: 48.5…50.5 °C
MDL Number: MFCD00015478
CAS Number: 79544-29-9
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR