4-Chloro-3-iodophenol ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

B25547.09 B25547.03
B25547.09EA 120 GBP
B25547.09 B25547.03
4-Chloro-3-iodophenol ≥98%
4-Chloro-3-iodophenol

Purity: 98% CAS No.: 202982-72-7 Molecular Formula: C6H4ClIO Formula Weight: 254.45 Melting Point: 75-77° MDL No.: MFCD00070769

Formula: C₆H₄ClIO
MW: 254.45 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00070769
CAS Number: 202982-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR