2-Chloro-4-iodo-5-nitrotoluene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2940-5G HC-2940-1G
COBBHC-2940-5GEA 0 GBP
COBBHC-2940-5G COBBHC-2940-1G
2-Chloro-4-iodo-5-nitrotoluene
2-Chloro-4-iodo-5-nitrotoluene
Formula: C₇H₅ClINO₂
MW: 297.48 g/mol
MDL Number: MFCD15143546
CAS Number: 859794-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR