2-Chloro-6-fluorophenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

OS-7579-5G OS-7579-25G OS-7579-100G
COBBOS-7579-5GEA 7.91 GBP
COBBOS-7579-5G COBBOS-7579-25G COBBOS-7579-100G
2-Chloro-6-fluorophenol
2-Chloro-6-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 199 °C (1013 hPa)
Melting Pt: 65 °C
MDL Number: MFCD01631574
CAS Number: 2040-90-6
EINECS: 433-890-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR