4-Bromo-2-nitroanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-1-methoxy-2-nitrobenzene

AN-1262-25G AN-1262-5G
COBBAN-1262-25GEA 0 GBP
COBBAN-1262-25G COBBAN-1262-5G
4-Bromo-2-nitroanisole
4-Bromo-2-nitroanisole
Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD00055529
CAS Number: 33696-00-3
EINECS: 251-642-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR