1-Bromo-4-hexylbenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Bromophenyl)hexane

OS-7603-5G OS-7603-25G OS-7603-1G
COBBOS-7603-5GEA 0 GBP
COBBOS-7603-5G COBBOS-7603-25G COBBOS-7603-1G
1-Bromo-4-hexylbenzene
1-Bromo-4-hexylbenzene
Formula: C₁₂H₁₇Br
MW: 241.17 g/mol
MDL Number: MFCD00061114
CAS Number: 23703-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR