1-Bromo-4-fluoro-2-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-5-fluoronitrobenzene

OS-7444-5G OS-7444-25G OS-7444-100G
COBBOS-7444-5GEA 0 GBP
COBBOS-7444-5G COBBOS-7444-25G COBBOS-7444-100G
1-Bromo-4-fluoro-2-nitrobenzene
1-Bromo-4-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD00055530
CAS Number: 446-09-3
EINECS: 207-160-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR