4-Bromo-2-fluorobenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009059-25G 009059-100G
FLUO009059-25GEA 0 GBP
FLUO009059-25G FLUO009059-100G
4-Bromo-2-fluorobenzonitrile
4-Bromo-2-fluorobenzonitrile
Formula: C₇H₃BrFN
Boiling Pt: 238…239 °C (1013 hPa)
Melting Pt: 71…73 °C
MDL Number: MFCD00143264
CAS Number: 105942-08-3
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR