4-Bromo-2,6-difluorobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
013773-1G
013773-5G
FLUO013773-1GEA
0
GBP
FLUO013773-1G
FLUO013773-5G
4-Bromo-2,6-difluorobenzonitrile
4-Bromo-2,6-difluorobenzonitrile
Formula:
C₇H₂BrF₂N MW: 218 g/mol |
MDL Number:
MFCD01310981 CAS Number: 123843-67-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...