4-Bromo-2-fluorobiphenyl

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-fluoro-1,1'-biphenyl

BK-4522-5G BK-4522-25G BK-4522-100G
COBBBK-4522-5GEA 0 GBP
COBBBK-4522-5G COBBBK-4522-25G COBBBK-4522-100G
4-Bromo-2-fluorobiphenyl
4-Bromo-2-fluorobiphenyl
Formula: C₁₂H₈BrF
MW: 251.1 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 38.5 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00051716
CAS Number: 41604-19-7
EINECS: 255-453-9
UN: 3152
ADR: 9,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR