1-Bromo-3-fluoro-2-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-6-fluoronitrobenzene

AN-1323-5G AN-1323-1G
COBBAN-1323-5GEA 0 GBP
COBBAN-1323-5G COBBAN-1323-1G
1-Bromo-3-fluoro-2-nitrobenzene
1-Bromo-3-fluoro-2-nitrobenzene
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
MDL Number: MFCD07368788
CAS Number: 886762-70-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR