4-Bromo-2-chloro-6-fluorophenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005575-1G 005575-5G
FLUO005575-1GEA 0 GBP
FLUO005575-1G FLUO005575-5G
4-Bromo-2-chloro-6-fluorophenol
4-Bromo-2-chloro-6-fluorophenol
Formula: C₆H₃BrClFO
MW: 225.44 g/mol
MDL Number: MFCD01631476
CAS Number: 161045-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR