2-Bromo-4,6-difluoroanisole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OT-1008-25G
OT-1008-1G
OT-1008-5G
COBBOT-1008-25GEA
0
GBP
COBBOT-1008-25G
COBBOT-1008-1G
COBBOT-1008-5G
2-Bromo-4,6-difluoroanisole
2-Bromo-4,6-difluoroanisole
Formula:
C₇H₅BrF₂O MW: 223.02 g/mol |
MDL Number:
MFCD00070754 CAS Number: 202865-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...