1-Bromo-3-(difluoromethoxy)benzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7695-5G OS-7695-1G OS-7695-25G
COBBOS-7695-5GEA 0 GBP
COBBOS-7695-5G COBBOS-7695-1G COBBOS-7695-25G
1-Bromo-3-(difluoromethoxy)benzene
1-Bromo-3-(difluoromethoxy)benzene
Formula: C₇H₅BrF₂O
MW: 223.02 g/mol
MDL Number: MFCD00236233
CAS Number: 262587-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR