1-Bromo-2-methoxynaphthalene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OT-1105-5G
OT-1105-1G
OT-1105-25G
COBBOT-1105-5GEA
0
GBP
COBBOT-1105-5G
COBBOT-1105-1G
COBBOT-1105-25G
1-Bromo-2-methoxynaphthalene
1-Bromo-2-methoxynaphthalene
Formula:
C₁₁H₉BrO MW: 237.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00055374 CAS Number: 3401-47-6 EINECS: 222-272-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...