4-Bromo-3-fluorophenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BK-4525-25G BK-4525-100G
COBBBK-4525-25GEA 0 GBP
COBBBK-4525-25G COBBBK-4525-100G
4-Bromo-3-fluorophenol
4-Bromo-3-fluorophenol
Formula: C₆H₄BrFO
MDL Number: MFCD00051907
CAS Number: 121219-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR