2-Bromo-6-fluorobenzonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4735-5G CA-4735-25G CA-4735-100G
COBBCA-4735-5GEA 0 GBP
COBBCA-4735-5G COBBCA-4735-25G COBBCA-4735-100G
2-Bromo-6-fluorobenzonitrile
2-Bromo-6-fluorobenzonitrile
Formula: C₇H₃BrFN
MW: 200.01 g/mol
MDL Number: MFCD06797753
CAS Number: 79544-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR