4-Bromo-3-chloro-2-fluoroiodobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-2-chloro-3-fluoro-4-iodobenzene

HC-3197-5G HC-3197-1G HC-3197-25G
COBBHC-3197-5GEA 0 GBP
COBBHC-3197-5G COBBHC-3197-1G COBBHC-3197-25G
4-Bromo-3-chloro-2-fluoroiodobenzene
4-Bromo-3-chloro-2-fluoroiodobenzene
Formula: C₆H₂BrClFI
MW: 335.34 g/mol
MDL Number: MFCD09878167
CAS Number: 1000573-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR