1-Bromo-2,5-difluoro-4-nitrobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Brom-2,5-difluor-4-nitrobenzol, 4-Bromo-2,5-difluoronitrobenzene

005502-1G 005502-5G
FLUO005502-1GEA 0 GBP
FLUO005502-1G FLUO005502-5G
1-Bromo-2,5-difluoro-4-nitrobenzene
1-Bromo-2,5-difluoro-4-nitrobenzene
Formula: C₆H₂BrF₂NO₂
MW: 237.99 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD00042184
CAS Number: 167415-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR