4-Bromo-3-fluoroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AN-1327-25G
AN-1327-100G
AN-1327-5G
COBBAN-1327-25GEA
0
GBP
COBBAN-1327-25G
COBBAN-1327-100G
COBBAN-1327-5G
4-Bromo-3-fluoroaniline
4-Bromo-3-fluoroaniline
Formula:
C₆H₅BrFN MW: 190.01 g/mol Boiling Pt: 231 °C (1013 hPa) Melting Pt: 72…73 °C |
MDL Number:
MFCD00672933 CAS Number: 656-65-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...