3-Methoxy-p-toluidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Amino-3-methoxy-4-methylbenzene, 4-Amino-2-methoxytoluene, 3-Methoxy-4-methylaniline, 4-Methyl-m-anisidine, 5-Amino-2-methylanisole

AN-1422-5G AN-1422-1G
COBBAN-1422-5GEA 0 GBP
COBBAN-1422-5G COBBAN-1422-1G
3-Methoxy-p-toluidine
3-Methoxy-p-toluidine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 250 °C (1013 hPa)
Melting Pt: 58 °C
MDL Number: MFCD00025371
CAS Number: 16452-01-0
EINECS: 240-500-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR