5-Chloro-2-iodoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-3130-25G HC-3130-5G
COBBHC-3130-25GEA 0 GBP
COBBHC-3130-25G COBBHC-3130-5G
5-Chloro-2-iodoaniline
5-Chloro-2-iodoaniline
Formula: C₆H₅ClIN
MW: 253.47 g/mol
MDL Number: MFCD06738960
CAS Number: 6828-35-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR