5-Bromo-o-anisidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromo-2-anisidine, 5-Bromo-2-methoxyaniline, 2-Amino-4-bromoanisole

AN-1281-5G AN-1281-25G AN-1281-1G
COBBAN-1281-5GEA 0 GBP
COBBAN-1281-5G COBBAN-1281-25G COBBAN-1281-1G
5-Bromo-o-anisidine
5-Bromo-o-anisidine
Formula: C₇H₈BrNO
MW: 202.05 g/mol
MDL Number: MFCD04037882
CAS Number: 6358-77-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR