2-Chloro-4,6-dibromoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-Chloro-2,4-dibromoaniline, 2,4-Dibromo-6-chloroaniline

AN-1566-100G AN-1566-25G AN-1566-5G
COBBAN-1566-100GEA 0 GBP
COBBAN-1566-100G COBBAN-1566-25G COBBAN-1566-5G
2-Chloro-4,6-dibromoaniline
2-Chloro-4,6-dibromoaniline
Formula: C₆H₄Br₂ClN
MW: 285.37 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 103 °C
Density: 2,305 g/cm³ (20 °C)
MDL Number: MFCD00031419
CAS Number: 874-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR