2-Bromo-4-chloroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AN-1188-5G
AN-1188-25G
AN-1188-100G
COBBAN-1188-5GEA
0
GBP
COBBAN-1188-5G
COBBAN-1188-25G
COBBAN-1188-100G
2-Bromo-4-chloroaniline
2-Bromo-4-chloroaniline
Formula:
C₆H₅BrClN MW: 206.47 g/mol Boiling Pt: 127 °C (20 mmHg) Melting Pt: 69 °C |
MDL Number:
MFCD00041313 CAS Number: 873-38-1 EINECS: 212-837-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...