2,6-Dibromoaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7645-25G OS-7645-100G OS-7645-5G
COBBOS-7645-25GEA 0 GBP
COBBOS-7645-25G COBBOS-7645-100G COBBOS-7645-5G
2,6-Dibromoaniline
2,6-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 85…86 °C
Storage Temperature: Ambient
MDL Number: MFCD00007638
CAS Number: 608-30-0
EINECS: 210-158-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR