2,6-Dibromo-4-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AN-1328-25G AN-1328-100G
COBBAN-1328-25GEA 0 GBP
COBBAN-1328-25G COBBAN-1328-100G
2,6-Dibromo-4-nitroaniline
2,6-Dibromo-4-nitroaniline
Formula: C₆H₄Br₂N₂O₂
MW: 295.92 g/mol
Melting Pt: 204…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00007639
CAS Number: 827-94-1
EINECS: 212-577-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR