Hydrocortisone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

SIALH0135-1MGEA 113 GBP
SIALH0135-1MG
Hydrocortisone, Sigma-Aldrich®
Hydrocortisone
Formula: C₂₁H₃₀O₅
MW: 362.47 g/mol
Boiling Pt: 567 °C (1013 hPa)
Melting Pt: 202…204 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00011654
CAS Number: 50-23-7
EINECS: 200-020-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR