1,3-Dimethoxy-4-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4-Dimethoxynitrobenzene, 2,4-Dimethoxy-1-nitrobenzene

AN-1348-5G AN-1348-1G
COBBAN-1348-5GEA 0 GBP
COBBAN-1348-5G COBBAN-1348-1G
1,3-Dimethoxy-4-nitrobenzene
1,3-Dimethoxy-4-nitrobenzene
Formula: C₈H₉NO₄
MW: 183.16 g/mol
Melting Pt: 72…76 °C
Density: 1.1876 g/cm³ (132 °C)
MDL Number: MFCD00024210
CAS Number: 4920-84-7
EINECS: 225-551-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR