1,3-Dibromo-5-nitrobenzene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1143-5G AN-1143-1G
COBBAN-1143-5GEA 0 GBP
COBBAN-1143-5G COBBAN-1143-1G
1,3-Dibromo-5-nitrobenzene
1,3-Dibromo-5-nitrobenzene
Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
MDL Number: MFCD00092529
CAS Number: 6311-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR