2,6-Dibromo-4-nitrophenol

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,3-Dibromo-2-hydroxy-5-nitrobenzene

OT-0697-25G OT-0697-5G OT-0697-100G
COBBOT-0697-25GEA 0 GBP
COBBOT-0697-25G COBBOT-0697-5G COBBOT-0697-100G
2,6-Dibromo-4-nitrophenol
2,6-Dibromo-4-nitrophenol
Formula: C₆H₃Br₂NO₃
Melting Pt: 148,5 °C
MDL Number: MFCD00007334
CAS Number: 99-28-5
EINECS: 202-744-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR